Order Entry
Export
ContactUsLinkComponent
Quin 2 potassium salt hydrate ≥95%
Irritant
Quin 2 potassium salt hydrate ≥95%
Catalog # AATB21052
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Quin 2 potassium salt hydrate ≥95%
Catalog # AATB21052
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :559.52 g/mol
ADR:5,II
 :GHS07
CAS Number:149022-19-5
Formula:C₂₆H₂₇N₃O
MDL Number:MFCD00167201