Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
EGTA, AM
EGTA, AM 10 mM calcium chelator
EGTA, AM 10 mM calcium chelator
AATB21006
:
AAT BIOQUEST
:
EGTA, AM 10 mM calcium chelator
AATB21006
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 mL
Seq:
2070
:
:
668.598 g/mol
:
5,II
:
GHS07
:
99590-86-0
:
C₂₆H₄₀N₂O₁₈