BAPTA tetrasodium salt
Catalog # AATB21004
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:564.37 g/mol
ADR:5,II
:GHS07
CAS Number:126824-24-6
Formula:C₂₂H₂₄N₂O₁₀
MDL Number:MFCD00067691