Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calbryte™ 630 potassium salt
Calbryte™ 630 potassium salt
Calbryte™ 630 potassium salt
Calbryte™ 630 potassium salt
AATB20727
:
AAT BIOQUEST
:
Calbryte™ 630 potassium salt
AATB20727
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
250 µG
Seq:
1370
:
:
5,II
:
1091.06 g/mol
:
GHS07