Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calbryte™ 520 potassium salt
Calbryte™ 520 potassium salt
Calbryte™ 520 potassium salt
:
AAT BIOQUEST
Calbryte™ 520 potassium salt
:
AAT BIOQUEST
:
:
5,II
:
909.02 g/mol
:
GHS07