Order Entry
Export
ContactUsLinkComponent
Fura-8FF™ AM
Irritant
Fura-8FF™ AM
Catalog # AATB20620
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Fura-8FF™ AM
Catalog # AATB20620
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 µG
  • Seq:
    1418

Specifications

About this item

 :
 :973.86 g/mol
ADR:5,II
 :GHS07