Order Entry
Export
ContactUsLinkComponent
Fluo-5N pentapotassium Salt Cell impermeant
Irritant
Fluo-5N pentapotassium Salt Cell impermeant
Catalog # AATB20567
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Fluo-5N pentapotassium Salt Cell impermeant
Catalog # AATB20567
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 µG
  • Seq:
    1418

Specifications

About this item

 :
 :958.06 g/mol
ADR:5,II
 :GHS07