Order Entry
Export
ContactUsLinkComponent
Cal-630™-Dextran conjugate, M.W. 10,000
Cal-630™-Dextran conjugate, M.W. 10,000
Catalog # AATB20546
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cal-630™-Dextran conjugate, M.W. 10,000
Catalog # AATB20546
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :11000 g/mol
ADR:5,II
 :GHS07