Cal-630™ AM
Supplier: AAT Bioquest
Jump to:
About this item
Calcium measurement is critical for numerous biological investigations.
- Improved sensitivity
- Improved calcium response
- Long-wavelength emission
- Cell permeant
Caution: For research use only.
- Molecular Weight:1282.89 g/mol
- ADR:5,II
3 Options Available Below
Product Details & Documents
Calcium measurement is critical for numerous biological investigations.
- Improved sensitivity
- Improved calcium response
- Long-wavelength emission
- Cell permeant
Caution: For research use only.
- Molecular Weight:1282.89 g/mol
- ADR:5,II



