Order Entry
Export
ContactUsLinkComponent
DNP maleimide FRET quencher precursor
Irritant
DNP maleimide FRET quencher precursor
Catalog # AATB2026
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
DNP maleimide FRET quencher precursor
Catalog # AATB2026
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :306.23 g/mol
ADR:5,II
 :GHS07
CAS Number:62024-74-2