0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
XFD350 C5 Maleimide
XFD350 C5 Maleimide fluorescent probe
XFD350 C5 Maleimide fluorescent probe
XFD350 C5 Maleimide fluorescent probe
Catalog # AATB1870
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
XFD350 C5 Maleimide fluorescent probe
Catalog # AATB1870
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
578.68 g/mol
ADR:
5,II
:
GHS07
Product Details