Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
XFD350 NHS Ester
XFD350 NHS Ester fluorescent probe
XFD350 NHS Ester fluorescent probe
XFD350 NHS Ester fluorescent probe
AATB1800
:
AAT BIOQUEST
:
XFD350 NHS Ester fluorescent probe
AATB1800
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
410.35 g/mol
:
5,II
:
GHS07