Order Entry
Export
ContactUsLinkComponent
Thiazole orange 10 mM in DMSO ≥90%
Thiazole orange 10 mM in DMSO ≥90%
Catalog # AATB17519
Supplier:  AAT Bioquest
CAS Number:  
undefined
Thiazole orange 10 mM in DMSO ≥90%
Catalog # AATB17519
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :476.61 g/mol
CAS Number:107091-89-4
Formula:C₂₆H₂₄N₂O₃S₂
MDL Number:MFCD00192499