
5-Propargylamino-3'-azidomethyl-dUTP
Catalog # AATB17093
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:50 Nmo
- Seq:9030
Specifications
About this item
:
:576.24 g/mol
ADR:5,II
:GHS07
CAS Number: 666847-57-0
Formula:C13H19N6O14P3