Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyanine 5 alkyne
Cyanine 5 alkyne ≥95% [equivalent to Cy5® alkyne]
Cyanine 5 alkyne ≥95% [equivalent to Cy5® alkyne]
:
AAT BIOQUEST
Cyanine 5 alkyne ≥95% [equivalent to Cy5® alkyne]
:
AAT BIOQUEST
:
:
807.9 g/mol