0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
TF4-VAD-FMK
TF4-VAD-FMK
TF4-VAD-FMK
AATB13471
:
AAT Bioquest
:
TF4-VAD-FMK
AATB13471
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
25 Tests
Seq:
0770
:
:
5,II
:
GHS07