(Ac-PAL)2R110
Plastic bottle, 1 mg
AATB13467
Jump to:
The non-fluorescent R110 substrates generate the bright green fluorescent rhodamine 110 product that has Ex/Em = 494/521 nm, and can be easily detected with a FITC filter set.
- Protease substrate
- Bright signal
- Multiple applications
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:1 mg