Order Entry
Export
Orders LinkContactUsLinkComponent
Z-LEHD-ProRed™ 620

Z-LEHD-ProRed™ 620

Plastic bottle, 1 mg
Catalog # AATB13435
Irritant
Supplier:   AAT Bioquest
Z-LEHD-ProRed™ 620
Catalog # AATB13435
Supplier:  AAT Bioquest
CAS Number:  
Z-LEHD-ProRed™ 620
Z-LEHD-ProRed™ 620

Plastic bottle, 1 mg

CAS Number:  
Price
469,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

ProRed™-derived protease substrates are colorless and non-fluorescent.

  • Caspase detection
  • Bright signal
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications