Order Entry
Export
ContactUsLinkComponent
 
Irritant
Ac-Leu-Glu-His-Asp-AMC ≥95%
Catalog # AATB13426
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Ac-Leu-Glu-His-Asp-AMC ≥95%
Catalog # AATB13426
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
ADR:5,II
 :711.729 g/mol
 :GHS07
CAS Number:292633-16-0
Formula:C₃₃H₄₁N₇O₁₁
MDL Number:MFCD03788104