Order Entry
Export
ContactUsLinkComponent
Beta-Ala-R110
Irritant
Beta-Ala-R110
Catalog # AATB13203
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Beta-Ala-R110
Catalog # AATB13203
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
ADR:5,II
 :GHS07