Order Entry
Export
ContactUsLinkComponent
D-Luciferin methyl ester ≥90%
Irritant
D-Luciferin methyl ester ≥90%
Catalog # AATB12514
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
D-Luciferin methyl ester ≥90%
Catalog # AATB12514
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :294.35 g/mol
ADR:5,II
 :GHS07
CAS Number:73918-26-0