Order Entry
Export
Orders LinkContactUsLinkComponent
PhosLite™ Green

PhosLite™ Green

Plastic bottle, 1 mg
Catalog # AATB11630
Irritant
Supplier:   AAT Bioquest
PhosLite™ Green
Catalog # AATB11630
Supplier:  AAT Bioquest
CAS Number:  
PhosLite™ Green
PhosLite™ Green

Plastic bottle, 1 mg

CAS Number:  
Price
313,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

This water-soluble PhosLite™ Green substrate generates a bright and photostable yellow-green fluorescent precipitate at the site of phosphtase activity.

  • Extremely large Stokes shift
  • Bright and stable signal
  • High photostability
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications