Order Entry
Export
Orders LinkContactUsLinkComponent
iFluor® 350, Hydrazide

iFluor® 350, Hydrazide

Plastic bottle, 1 mg
Catalog # AATB1080
Irritant
Supplier:   AAT Bioquest
iFluor® 350, Hydrazide
Catalog # AATB1080
Supplier:  AAT Bioquest
CAS Number:  
iFluor® 350, Hydrazide
iFluor® 350, Hydrazide

Plastic bottle, 1 mg

CAS Number:  
Price
313,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.

  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications