Order Entry
Export
Orders LinkContactUsLinkComponent
iFluor® 350, Amine

iFluor® 350, Amine

Plastic bottle, 1 mg
Catalog # AATB1070
Irritant
Supplier:   AAT Bioquest
iFluor® 350, Amine
Catalog # AATB1070
Supplier:  AAT Bioquest
CAS Number:  
iFluor® 350, Amine
iFluor® 350, Amine

Plastic bottle, 1 mg

CAS Number:  
Price
313,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

iFluor® dyes are optimized for labeling proteins, in particular, antibodies.

  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications