0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
iFluor®
iFluor® 430, Maleimide
iFluor® 430, Maleimide
AATB1054
:
AAT BIOQUEST
:
iFluor® 430, Maleimide
AATB1054
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
5,II
:
GHS07