Carbendazim, colourless, liquid
Catalog # EHERA10990000ME100
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:1 mL
- Seq:2070
Specifications
Specification Test Results
- Autoignition temperature:464 °C
- Dynamic viscosity:0.544 - 0.59 mPa s
- Partition coefficient:−0.77
- Vapour pressure:128 hPa
- Relative density:0.791
- Relative vapour density:1.1
- Stability:Stable under normal conditions
- Sensitivity to static discharge:Yes
Specification Test Results
About this item
:
:191.19 g/mol
Boiling Pt:64.7 °C
Melting Pt:−98 °C
Melting Pt:–98 °C
Flash Pt:11 °C
UN:1230
ADR:3,II
:GHS02
:GHS06
:GHS08
CAS Number:10605-21-7
Formula:C₉H₉N₃O₂
MDL Number:MFCD00055390