Aluminized Heat Protection Coats
Supplier: JUTEC
Certificates
About this item
Coat made of preox-aramide fabric heat protection coat.
- Flexible fabric
- Very pleasant wearing comfort
- High heat reflection
- Insulated snaps, stand-up collar, Velcro fastener
- Weight: 260 g/m²
Certifications: EN ISO 11612:2015.
Specifications
- Catalog No:
- JUTEHSM120KA-1-56
- JUTEHSM120KA-1-54
- Garment application:
- Industrial
- Industrial
- Risk area:
- Heat
- Heat
- Sleeve or leg length:
- Long
- Long