VWR® Viscometers
Supplier: VWR Collection
Jump to:
About this item
The Cannon-Fenske routine viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104.
Minimum sample volume is 11 ml. Minimum bath depth is 241 mm (9,5")
2 Options Available Below
Product Details & Documents
The Cannon-Fenske routine viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104.
Minimum sample volume is 11 ml. Minimum bath depth is 241 mm (9,5")
Product Family Options
Product Information
- RangeConstantAvailabilityPrice
- VWRU10791-456Calibrated, size: 0C
- VWRU10791-458Calibrated, size: 0B