Orders LinkContactUsLinkComponent
Filter Fleece
Filter Fleece
Filter fleece
Catalog # INFIPPV75-37-100
Supplier:   PIEPER FILTER
Filter Fleece
Catalog # INFIPPV75-37-100
Supplier:  PIEPER FILTER
Filter Fleece
Filter fleece
Price
429,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Filter fleece is suitable for drainage under membranes. It is also used in chipping processes to keep the coolant free of the chips. The fleece is also suitable for coarse air filtration.
  • Polypropylene fleece
  • Thickness: 75 µm
  • Sampling according to IFA 7512, IFA 6172
Specifications
  • Description:
    Filter fleece