ContactUsLinkComponent
FlashPure EcoFlex Silica Cartridges
 :  BUCHI
FlashPure EcoFlex Silica Cartridges
 :  BUCHI
  • Catalog No:
  • 140000027.
  • 140000028.
  • 140000025.
  • 140000026.
  • 140000029.
  • 140000030.
  • 140000031.
  • 140000023.
  • 140000024.
  • 140000021.
  • 11072741.
  • 140000022.
  • Phase name:
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • FlashPure EcoFlex silica
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Max. pressure:
  • 10 bar
  • 10 bar
  • 14 bar
  • 14 bar
  • 7 bar
  • 7 bar
  • 7 bar
  • 20 bar
  • 20 bar
  • 20 bar
  • 7 bar
  • 20 bar
  • Surface area:
  • ≥480 m²/g
  • ≥480 m²/g
  • ≥480 m²/g
  • ≥480 m²/g
  • ≥480 m²/g
  • ≥480 m²/g
  • ≥480 m²/g
  • ≥480 m²/g
  • ≥480 m²/g
  • ≥480 m²/g
  • ≥480 m²/g
  • ≥480 m²/g
  • Polarity:
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase