Cetuximab (Human) ELISA kit
ABNOKA6822.1KIT
: Abnova
:
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:Cetuximab
- Assay Principle:Quantitative
Cetuximab (Human) Elisa Kit (Quantitative) is a quantitative determination of free Cetuximab in serum and plasma.