Golimumab (Human) ELISA kit
Catalog # ABNOKA6803.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:Golimumab
- Assay Principle:Quantitative
Specifications
About this item
Golimumab (Human) ELISA Kit (Quantitative) is a quantitative determination of free golimumab in serum and plasma.