The Albumin (Monkey) ELISA kit
Catalog # ABNOKA6757.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Monkey
- Target protein:Albumin
- Assay Principle:Quantitative
Specifications
About this item
The Albumin (Monkey) ELISA Kit is used for quantitative determination of albumin in monkey plasma and serum samples.