Order Entry
Export
ContactUsLinkComponent
 
IgG2a (Rat) ELISA kit
  ABNOKA6759.1KIT
 :  Abnova
 :  
IgG2a (Rat) ELISA kit
  ABNOKA6759.1KIT
 :  Abnova
 :  

 

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Rat
  • Target protein:
    IgG2a
  • Assay Principle:
    Quantitative

 

 

IgG2a (Rat) ELISA Kit is for the quantitative measurement of rat IgG2a in serum or plasma.