Order Entry
Export
ContactUsLinkComponent
IgA (Goat) ELISA kit
IgA (Goat) ELISA kit
Catalog # ABNOKA6767.1KIT
Supplier:  Abnova
CAS Number:  
IgA (Goat) ELISA kit
Catalog # ABNOKA6767.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Goat
  • Target protein:
    IgA
  • Assay Principle:
    Quantitative

Specifications

About this item

IgA (Goat) ELISA Kit is for the quantitative measurement of goat IgA in serum or plasma.