Order Entry
Export
ContactUsLinkComponent
 
The F8 (Human) ELISA kit
  ABNOKA6758.1KIT
 :  Abnova
 :  
The F8 (Human) ELISA kit
  ABNOKA6758.1KIT
 :  Abnova
 :  

 

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    F8
  • Assay Principle:
    Quantitative

 

 

The F8 (Human) ELISA Kit is used for quantitative determination of Human F8 in serum and plasma.