Mumps Virus IgG (Human) ELISA kit
Catalog # ABNOKA6783.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:IgG
- Assay Principle:Qualitative
Specifications
About this item
Mumps Virus IgG (Human) ELISA Kit is a qualitative determination of IgG class antibodies against Mumps Virus in human serum or plasma.