KLH IgG (Human) ELISA kit
Catalog # ABNOKA6737.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:IgG
- Assay Principle:Quantitative
Specifications
About this item
KLH IgG (Human) ELISA Kit is for the quantitative measurement of human anti-KLH IgG in serum, plasma and other fluids.