Bordetella pertussis IgG ELISA kit
Catalog # ABNOKA6719.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:B. pertussis
- Target protein:IgG
- Assay Principle:Qualitative
Specifications
About this item
Bordetella pertussis IgG ELISA Kit is a qualitative determination of IgM class antibodies against Bordetella pertussis in human serum or plasma.