Order Entry
Export
ContactUsLinkComponent
 
Progesterone (Human) ELISA kit
Catalog # ABNOKA6703.1KIT
Supplier:  Abnova
CAS Number:  
Progesterone (Human) ELISA kit
Catalog # ABNOKA6703.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    Progesterone
  • Assay Principle:
    Quantitative

Specifications

About this item

Progesterone (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of progesterone in serum and plasma.