Free beta-CG (Human) ELISA kit
Catalog # ABNOKA6711.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:Free beta-CG
- Assay Principle:Quantitative
Specifications
About this item
Free beta-CG (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of free beta-chorionic gonadotropin in serum and plasma.