Order Entry
Export
Orders LinkContactUsLinkComponent
 

3-Nitrotyrosine ELISA kit

1 KIT
Catalog # ABNOKA6682.1KIT
Supplier:   Abnova
3-Nitrotyrosine ELISA kit
Catalog # ABNOKA6682.1KIT
Supplier:  Abnova
CAS Number:  
3-Nitrotyrosine ELISA kit

1 KIT

CAS Number:  
Price
573,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

3-Nitrotyrosine ELISA Kit is a quantitative determination of 3-NT concentrations in serum, plasma and other biological fluids.

Specifications

Specifications

  • Pk:
    1 KIT
  • Cat. No.:
    ABNOKA6682.1KIT
  • Target protein:
    3-Nitrotyrosine
  • Assay Principle:
    Quantitative

Specifications