Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human CD22hFc tag
Recombinant Human CD22hFc tag
Recombinant Human CD22hFc tag
ABNOP7654.100UG
:
Abnova
:
Recombinant Human CD22hFc tag
ABNOP7654.100UG
:
Abnova
:
Pk:
100 µG
Protein/peptide type:
Recombinant
Species:
Human
Tag sequence:
hFc tag
Gene ID:
P20273
Protein/peptide name:
CD22
Sequence:
20a.a.-687a.a.