Order Entry
Export
ContactUsLinkComponent
Recombinant Human CCL19 (from E. coli), T7-tag
Recombinant Human CCL19 (from E. coli), T7-tag
Catalog # ABNOP9545.20UG
Supplier:  Abnova
CAS Number:  
Recombinant Human CCL19 (from E. coli), T7-tag
Catalog # ABNOP9545.20UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    T7-tag
  • Gene ID:
    Q99731
  • Protein/peptide name:
    CCL19
  • Sequence:
    22a.a.-98a.a.

Specifications

About this item