Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human GCDH (from E. coli), His-tag
Recombinant Human GCDH (from E. coli), His-tag
ABNOP9597.10UG
:
Abnova
:
Recombinant Human GCDH (from E. coli), His-tag
ABNOP9597.10UG
:
Abnova
:
Pk:
10 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Tag sequence:
His-tag
Gene ID:
Q92947
Protein/peptide name:
GCDH
Sequence:
45a.a.-438a.a.