Order Entry
Export
ContactUsLinkComponent
Recombinant Human TNFRSF21 (from Baculovirus), His-tag
Recombinant Human TNFRSF21 (from Baculovirus), His-tag
Catalog # ABNOP9278.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Human TNFRSF21 (from Baculovirus), His-tag
Catalog # ABNOP9278.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Protein/peptide name:
    TNFRSF21

Specifications

About this item