Order Entry
Export
ContactUsLinkComponent
Recombinant Human MANF (from E. coli)
Recombinant Human MANF (from E. coli)
Catalog # ABNOP8915.20UG
Supplier:  Abnova
CAS Number:  
Recombinant Human MANF (from E. coli)
Catalog # ABNOP8915.20UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    P55145
  • Protein/peptide name:
    MANF
  • Sequence:
    25a.a.-182a.a.

Specifications

About this item