Order Entry
Export
ContactUsLinkComponent
Recombinant Human CSF1 (from E. coli)
Recombinant Human CSF1 (from E. coli)
Catalog # ABNOP8922.25UG
Supplier:  Abnova
CAS Number:  
Recombinant Human CSF1 (from E. coli)
Catalog # ABNOP8922.25UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    P09603
  • Protein/peptide name:
    CSF1
  • Sequence:
    33a.a.-190a.a.

Specifications

About this item