Order Entry
Export
ContactUsLinkComponent
Recombinant Mouse NRP1 (from Baculovirus), His-tag
Recombinant Mouse NRP1 (from Baculovirus), His-tag
Catalog # ABNOP8968.5UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse NRP1 (from Baculovirus), His-tag
Catalog # ABNOP8968.5UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Mouse
  • Tag sequence:
    His-tag
  • Gene ID:
    P97333
  • Protein/peptide name:
    NRP1
  • Sequence:
    22a.a.-856a.a.

Specifications

About this item