Recombinant Human CXCL10 (from E. coli), His-tag
Catalog # ABNOP9490.25UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:25 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:P02778
- Protein/peptide name:CXCL10
- Sequence:22a.a.-98a.a.